Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZIM2 Full-length ORF (NP_056178.3, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967919 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-919 10 μg
89-967-920 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-919 Supplier Abnova™ Supplier No. H00023619P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5' exons and have a common promoter, and both genes are paternally expressed (PMID:15203203). Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Additional 5' alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene. [provided by RefSeq]

Sequence: MYQPEDDNNSDVTSDDDMTRNRRESSPPHSVHSFSGDRDWDRRGRSRDMEPRDRWSHTRNPRSRMPPRDLSLPVVAKTSFEMDREDDRDSRAYESRSQDAESYQNVVDLAEDRKPHNTIQDNMENYRKLLSLGFLAQDSVPAEKRNTEMLDNLPSAGSQFPDFKHLGTFLVFEELVTFEDVLVDFSPEELSSLSAAQRNLYREVMLENYRNLVSLGHQFSKPDIISRLEEEESYAMETDSRHTVICQGESHDDPLEPHQGNQEKLLTPITMNDPKTLTPERSYGSDEFERSSNLSKQSKDPLGKDPQEGTAPGICTSPQSASQENKHNRCEFCKRTFSTQVALRRHERIHTGKKPYECKQCAEAFYLMPHLNRHQKTHSGRKTSGCNEGRKPSVQCANLCERVRIHSQEDYFECFQCGKAFLQNVHLLQHLKAHEAARVLPPGLSHSKTYLIRYQRKHDYVGERACQCCDCGRVFSRNSYLIQHYRTHTQERPYQCQLCGKCFGRPSYLTQHYQLHSQEKTVECDHC

Specifications

Accession Number NP_056178.3
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23619
Molecular Weight (g/mol) 87.6kDa
Name ZIM2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MYQPEDDNNSDVTSDDDMTRNRRESSPPHSVHSFSGDRDWDRRGRSRDMEPRDRWSHTRNPRSRMPPRDLSLPVVAKTSFEMDREDDRDSRAYESRSQDAESYQNVVDLAEDRKPHNTIQDNMENYRKLLSLGFLAQDSVPAEKRNTEMLDNLPSAGSQFPDFKHLGTFLVFEELVTFEDVLVDFSPEELSSLSAAQRNLYREVMLENYRNLVSLGHQFSKPDIISRLEEEESYAMETDSRHTVICQGESHDDPLEPHQGNQEKLLTPITMNDPKTLTPERSYGSDEFERSSNLSKQSKDPLGKDPQEGTAPGICTSPQSASQENKHNRCEFCKRTFSTQVALRRHERIHTGKKPYECKQCAEAFYLMPHLNRHQKTHSGRKTSGCNEGRKPSVQCANLCERVRIHSQEDYFECFQCGKAFLQNVHLLQHLKAHEAARVLPPGLSHSKTYLIRYQRKHDYVGERACQCCDCGRVFSRNSYLIQHYRTHTQERPYQCQLCGKCFGRPSYLTQHYQLHSQEKTVECDHC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ZNF656
Common Name ZIM2
Gene Symbol ZIM2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.