Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ZMYM2 Synthetic Peptide

Catalog No. PEP1204
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1204 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1204 Supplier Invitrogen™ Supplier No. PEP1204
Add to Cart
Add to Cart

Synthetic Peptide

ZMYM2 Synthetic Peptide, PEP-1204, from Invitrogen.

ZNF198 is a ubiquitously expressed nuclear protein whose physiological role is largely unknown. It consists of 5 N-terminal zinc finger motifs responsible for protein-protein interactions, a central proline-rich domain and C-terminal nuclear localization signal (NLS) and an acidic domain. Reciprocal chromosomal translocation involving ZNF198 and FGFR1 results in the formation of ZNF198/FGFR1 fusion kinase protein. This fusion protein is a ligand-dependent, constitutively active cytoplasmic tyrosine kinase and regulates several STAT transcription factors including STAT 1, 3 and 5. This fusion protein is an oncogenic protein and is associated with an atypical myeloproliferative disease, peripheral blood eosinophilia and T-cell leukemia/lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7750
Species Human
Purification Method purified
Accession Number Q9UBW7
Common Name Human ZMYM2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol ZMYM2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.