Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF10 Full-length ORF (NP_056209.2, 1 a.a. - 573 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89967960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-960 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-960 Supplier Abnova™ Supplier No. H00007556P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene contains a C2H2 zinc finger, and has been shown to function as a transcriptional repressor. The Kruppel-associated box (KRAB) domain of this protein is found to be responsible for its transcriptional repression activity. RING finger containing protein TIF1 was reported to interact with the KRAB domain, and may serve as a mediator for the repression activity of this protein. [provided by RefSeq]

Sequence: MDAKSLTAWSRTLVTFKDVFVDFTREEWKLLDTAQQIVYRNVMLENYKNLVSLGYQLTKPDVILRLEKGEEPWLVEREIHQETHPDSETAFEIKSSVSSRSIFKDKQSCDIKMEGMARNDLWYLSLEEVWKCRDQLDKYQENPERHLRQVAFTQKKVLTQERVSESGKYGGNCLLPAQLVLREYFHKRDSHTKSLKHDLVLNGHQDSCASNSNECGQTFCQNIHLIQFARTHTGDKSYKCPDNDNSLTHGSSLGISKGIHREKPYECKECGKFFSWRSNLTRHQLIHTGEKPYECKECGKSFSRSSHLIGHQKTHTGEEPYECKECGKSFSWFSHLVTHQRTHTGDKLYTCNQCGKSFVHSSRLIRHQRTHTGEKPYECPECGKSFRQSTHLILHQRTHVRVRPYECNECGKSYSQRSHLVVHHRIHTGLKPFECKDCGKCFSRSSHLYSHQRTHTGEKPYECHDCGKSFSQSSALIVHQRIHTGEKPYECCQCGKAFIRKNDLIKHQRIHVGEETYKCNQCGIIFSQNSPFIVHQIAHTGEQFLTCNQCGTALVNTSNLIGYQTNHIRENAY

Specifications

Accession Number NP_056209.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7556
Molecular Weight (g/mol) 92.9kDa
Name ZNF10 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MDAKSLTAWSRTLVTFKDVFVDFTREEWKLLDTAQQIVYRNVMLENYKNLVSLGYQLTKPDVILRLEKGEEPWLVEREIHQETHPDSETAFEIKSSVSSRSIFKDKQSCDIKMEGMARNDLWYLSLEEVWKCRDQLDKYQENPERHLRQVAFTQKKVLTQERVSESGKYGGNCLLPAQLVLREYFHKRDSHTKSLKHDLVLNGHQDSCASNSNECGQTFCQNIHLIQFARTHTGDKSYKCPDNDNSLTHGSSLGISKGIHREKPYECKECGKFFSWRSNLTRHQLIHTGEKPYECKECGKSFSRSSHLIGHQKTHTGEEPYECKECGKSFSWFSHLVTHQRTHTGDKLYTCNQCGKSFVHSSRLIRHQRTHTGEKPYECPECGKSFRQSTHLILHQRTHVRVRPYECNECGKSYSQRSHLVVHHRIHTGLKPFECKDCGKCFSRSSHLYSHQRTHTGEKPYECHDCGKSFSQSSALIVHQRIHTGEKPYECCQCGKAFIRKNDLIKHQRIHVGEETYKCNQCGIIFSQNSPFIVHQIAHTGEQFLTCNQCGTALVNTSNLIGYQTNHIRENAY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KOX1
Common Name ZNF10
Gene Symbol ZNF10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.