Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF254 Full-length ORF (AAH43147.2, 1 a.a. - 659 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968116 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-116 25 μg
89-968-115 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-116 Supplier Abnova™ Supplier No. H00009534P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]

Sequence: MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV

Specifications

Accession Number AAH43147.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9534
Molecular Weight (g/mol) 103.5kDa
Name ZNF254 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MPGPPRSLEMGLLTFRDVAIEFSLEEWQHLDIAQQNLYRNVMLENYRNLAFLGIAVSKPDLITCLEQGKEPWNMKRHEMVDEPPGMCPHFAQGLWPEQGMEDSFQKAILRRYGKYGHENLQLRKGCKSVDEYKVNKEGYNGLNQCFTTAQSKVFQCDKYLKVFYKFLNSNRPKIRHTEKKSFKCKKRVKLFCMLSHKTQHKSIYHREKSYKCKECGKTFNWSSTLTNHRKIYTEEKPYKCEEYNKSPKQLSTLTTHEIIHAGEKLYKCEECGEAFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTEHKKIHTRKKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKAFSQSSTLTTHKIIHTGEKRYKCLECGKAFKQLSTLTTHKIIHVGEKLYKCEECGKGFNRSSNLTTHKIIHTGEKPYKCEECGKAFIWSSTLTKHKRIHTREKPYKCEECGKAFIWSSTLTRHKRMHTGEKPYKCEECGKSFSQSSTLTTHKIIHTGEKPYKCEECGKAFNWSSTLTKHNIIHTEEKPYKCEKCGKAFKQSSILTNHKRIHTGEKPYKCEECGKSFNRSSTFTKHKVIHTGVKPYKCEECGKAFFWSSTLTKHKRIHTGEQPYKWEKFGKAFNRSSHLTTDKITHWREILQV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BMZF-5/FLJ58216/HD-ZNF1/ZNF539/ZNF91L
Common Name ZNF254
Gene Symbol ZNF254
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.