Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF346 Full-length ORF (NP_036411.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968226 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-226 10 μg
89-968-227 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-226 Supplier Abnova™ Supplier No. H00023567P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a nucleolar, zinc finger protein that preferentially binds to double-stranded (ds) RNA or RNA/DNA hybrids, rather than DNA alone. Mutational studies indicate that the zinc finger domains are not only essential for dsRNA binding, but are also required for its nucleolar localization. The encoded protein may be involved in cell growth and survival. [provided by RefSeq]

Sequence: MEYPAPATVQAADGGAAGPYSSSELLEGQEPDGVRFDRERARRLWEAVSGAQPVGREEVEHMIQKNQCLFTNTQCKVCCALLISESQKLAHYQSKKHANKVKRYLAIHGMETLKGETKKLDSDQKSSRSKDKNQCCPICNMTFSSPVVAQSHYLGKTHAKNLKLKQQSTKVEALHQNREMIDPDKFCSLCHATFNDPVMAQQHYVGKKHRKQETKLKLMARYGRLADPAVTDFPAGKGYPCKTCKIVLNSIEQYQAHVSGFKHKNQSPKTVASSLGQIPMQRQPIQKDSTTLED

Specifications

Accession Number NP_036411.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23567
Molecular Weight (g/mol) 59.3kDa
Name ZNF346 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEYPAPATVQAADGGAAGPYSSSELLEGQEPDGVRFDRERARRLWEAVSGAQPVGREEVEHMIQKNQCLFTNTQCKVCCALLISESQKLAHYQSKKHANKVKRYLAIHGMETLKGETKKLDSDQKSSRSKDKNQCCPICNMTFSSPVVAQSHYLGKTHAKNLKLKQQSTKVEALHQNREMIDPDKFCSLCHATFNDPVMAQQHYVGKKHRKQETKLKLMARYGRLADPAVTDFPAGKGYPCKTCKIVLNSIEQYQAHVSGFKHKNQSPKTVASSLGQIPMQRQPIQKDSTTLED
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp547M223/JAZ/Zfp346
Common Name ZNF346
Gene Symbol ZNF346
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.