Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF684 Full-length ORF (NP_689586.2, 1 a.a. - 378 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968507 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-507 25 μg
89-968-506 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-507 Supplier Abnova™ Supplier No. H00127396P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sequence: MISFQESVTFQDVAVDFTAEEWQLLDCAERTLYWDVMLENYRNLISVGCPITKTKVILKVEQGQEPWMVEGANPHESSPESDYPLVDEPGKHRESKDNFLKSVLLTFNKILTMERIHHYNMSTSLNPMRKKSYKSFEKCLPPNLDLLKYNRSYTVENAYECSECGKAFKKKFHFIRHEKNHTRKKPFECNDCGKAYSRKAHLATHQKIHNGERPFVCNDCGKAFMHKAQLVVHQRLHTGEKPYECSQCGKTFTWNSSFNQHVKSHTLEKSFECKECGKTFRYSSSLYKHSRFHTGEKPYQCIICGKAFGNTSVLVTHQRIHTGEKPYSCIECGKAFIKKSHLLRHQITHTGEKPYECNRCGKAFSQKSNLIVHQKIHT

Specifications

Accession Number NP_689586.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 127396
Molecular Weight (g/mol) 70.3kDa
Name ZNF684 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MISFQESVTFQDVAVDFTAEEWQLLDCAERTLYWDVMLENYRNLISVGCPITKTKVILKVEQGQEPWMVEGANPHESSPESDYPLVDEPGKHRESKDNFLKSVLLTFNKILTMERIHHYNMSTSLNPMRKKSYKSFEKCLPPNLDLLKYNRSYTVENAYECSECGKAFKKKFHFIRHEKNHTRKKPFECNDCGKAYSRKAHLATHQKIHNGERPFVCNDCGKAFMHKAQLVVHQRLHTGEKPYECSQCGKTFTWNSSFNQHVKSHTLEKSFECKECGKTFRYSSSLYKHSRFHTGEKPYQCIICGKAFGNTSVLVTHQRIHTGEKPYSCIECGKAFIKKSHLLRHQITHTGEKPYECNRCGKAFSQKSNLIVHQKIHT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC27466
Common Name ZNF684
Gene Symbol ZNF684
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.