Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF83 Partial ORF (NP_060770, 1 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-596 25 μg
89-968-595 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-596 Supplier Abnova™ Supplier No. H00055769Q01L

Used for AP, Array, ELISA, WB-Re

Sequence: MHGRKDDAQKQPVKNQLGLNPQSHLPELQLFQAEGKIYKYDHMEKSVNSSSLVSPPQRISSTVKTHISHTYECNFVDSLFTQKEKANIGTEHYKCNERGKAFHQGLHFTIH

Specifications

Accession Number NP_060770
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55769
Molecular Weight (g/mol) 37.95kDa
Name ZNF83 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MHGRKDDAQKQPVKNQLGLNPQSHLPELQLFQAEGKIYKYDHMEKSVNSSSLVSPPQRISSTVKTHISHTYECNFVDSLFTQKEKANIGTEHYKCNERGKAFHQGLHFTIH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ11015/FLJ14876/FLJ30097/FLJ90585/HPF1/MGC33853/ZNF816B
Common Name ZNF83
Gene Symbol ZNF83
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.