Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNRF1 Partial ORF (NP_115644, 74 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968630 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-630 25 μg
89-968-629 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-630 Supplier Abnova™ Supplier No. H00084937Q01L

Used for AP, Array, ELISA, WB-Re

In a study identifying genes in rat that are upregulated in response to nerve damage, a gene which is highly expressed in ganglia and in the central nervous system was found. The protein encoded by the rat gene contains both a zinc finger and a RING finger motif and is localized in the endosome/lysosome compartment, indicating that it may be involved in ubiquitin-mediated protein modification. The protein encoded by this human gene is highly similar in sequence to that encoded by the rat gene. [provided by RefSeq]

Sequence: PASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRLSYNDDVLT

Specifications

Accession Number NP_115644
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84937
Molecular Weight (g/mol) 37.29kDa
Name ZNRF1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen PASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRLSYNDDVLT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434E229/FLJ14846/MGC15430/NIN283
Common Name ZNRF1
Gene Symbol ZNRF1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.