Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZP2 Full-length ORF (AAH96305.1, 1 a.a. - 736 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968645 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-645 25 μg
89-968-644 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-645 Supplier Abnova™ Supplier No. H00007783P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in secondary binding and penetration of acrosome-reacted spermatozoa. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a consensus furin cleavage site, and a C-terminal transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies. [provided by RefSeq]

Sequence: MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNH

Specifications

Accession Number AAH96305.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7783
Molecular Weight (g/mol) 107.7kDa
Name ZP2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ZPA
Common Name ZP2
Gene Symbol ZP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.