Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZRF1 Full-length ORF (AAH56682.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89968660 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-660 25 μg
89-968-659 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-968-660 Supplier Abnova™ Supplier No. H00027000P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the M-phase phosphoprotein (MPP) family. The gene encodes a phosphoprotein with a J domain and a Myb DNA-binding domain which localizes to both the nucleus and the cytosol. The protein is capable of forming a heterodimeric complex that associates with ribosomes, acting as a molecular chaperone for nascent polypeptide chains as they exit the ribosome. This protein was identified as a leukemia-associated antigen and expression of the gene is upregulated in leukemic blasts. Also, chromosomal aberrations involving this gene are associated with primary head and neck squamous cell tumors. This gene has a pseudogene on chromosome 6. Alternatively spliced variants which encode different protein isoforms have been described. [provided by RefSeq]

Sequence: MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQ

Specifications

Accession Number AAH56682.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27000
Molecular Weight (g/mol) 45.4kDa
Name ZRF1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MPHOSPH11/MPP11/ZRF1/ZUO1
Common Name DNAJC2
Gene Symbol DNAJC2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.