Learn More
Humboldt Single-Size-Opening Tripods
SDP
Iron ring is supported on three legs to eliminate rocking of support
$157.10 - $252.00
Specifications
Material Cast Iron
Product Type Flanged Concentric Single Ring
For Use With (Application) Iron ring is supported on three legs to eliminate rocking of support
Quantity 10 Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (English) Outer Diameter (English) Inner Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Outer Diameter (English) Inner Height (English) Price Quantity & Availability  
15-295
SDP
View Documents
Humboldt
H24133
3.5 in. 1.2 in. 6 in.
Each for $157.10
Only null left
Add to Cart
15-300C
SDP
View Documents
Humboldt
H24180
8 in. 6 in. 9 in.
Each for $252.00
Only null left
Add to Cart
Description
Description
  • Ring for 15-295 is without inside flange
  • 15-295 is 6 in. high (15cm); 15-300C, 9 in. (23cm)
Specifications
Specifications
Cast Iron
Flanged Concentric Single Ring
Iron ring is supported on three legs to eliminate rocking of support
10 Pk.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.