Learn More
Caron Products And Services Humidified Incubator, 33 cu. ft., Temp & RH
Supplier:  Caron Products And Services 7003332
Encompass_Preferred
33 ft3 (934 L) Humidified Incubator offers temperature (10C above ambient-70C) and humidity control (ambient to 98% RH). Ideal for a wide range of applications. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.
Catalog No. NC2676803
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.