Learn More
BRINSEA PRODUCTS Ova-Easy Advance Humidity Pump Rigid Tubing
Supplier:  BRINSEA PRODUCTS 14.440
Encompass_Preferred
Replacement rigid water tubing used with the Ova-Easy Advance Humidity Pump (part # NC1728213) which provides fully automatic control of humidity when paired with any of the Brinsea Ova-Easy Cabinet Incubators.
Catalog No. NC2184512
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.