Description
Includes
SCS traceable calibration certificate, textile pads, Calibration instructions
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
- Ampoules contain unsaturated salt solutions
- Simple and safe on-site calibration method
- Unlimited storage life
Specifications
Specifications
| Packaging | 0.8 |
| Certificate of Analysis | SCS traceable calibration certificate, textile pad, calibration instruction |
| Technique | On-site Calibration and Adjustment of Rotronic Probe |
| For Use With (Equipment) | Rotronic Instrument |
| Solution Type | 80% RH |
| Type | Humidity Standard |