Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Humidity and Temperature Data Logger
SDP

Catalog No. 15059306 Shop All Fisherbrand Products
Change view
Click to view available options
Certifications/Compliance:
CE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
15-059-306 CE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-059-306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Humidity/Temperature recorder. (Requires USB Interface Cable Package - sold separately, Cat. No. 15-060-036).

Ultra-high speed download capability

Using the data logging software, starting, stopping and downloading from the unit is simple and easy. Graphical, tabular and summary data is provided for analysis and data can be viewed in °C, °F, K,°R, %RH, mg/mL water vapor concentration and dew point. The data can also be automatically exported to Excel™ for further calculations. Free firmware upgrades for the life of the product are available, so that data loggers already deployed in the field can grow with the technology. Units do not need to be returned to the factory for upgrades. The user can do this automatically from any PC.

  • 10 year battery life
  • One-second reading rate
  • Multiple start/stop function
  • Ultra-high speed download capability
  • 500,000 reading per channel storage capacity
  • Optional memory wrap
  • Battery life indicator
  • Optional password protection
  • Programmable high-low alarms
Temperature
  • Temperature sensor: precision RTD element
  • Temperature range: -40°C to +80°C (-40°F to +176°F)
  • Temperature resolution: 0.01°C (0.018°F)
  • Temperature calibrated accuracy: ±0.5°C (±0.9°F)
Humidity
  • Humidity sensor: internal semiconductor
  • Humidity range: 0ã95% RH
  • Humidity resolution: 0.1% RH
  • Humidity calibrated accuracy: ±3.0%RH (±2%RH typical at 25°C/77°F)
  • RH Units: %RH, dew point, water vapor, concentration (mg/hl)
General
  • Memory: 500,000 readings per channel; software configurable memory wrap; 250,000 readings in multiple start/stop mode
  • Reading rate: 1 reading every second to 1 reading every 24 hours
  • Alarm: Programmable high and low limits; activated when humidity reaches or exceeds set limits
  • Baud rate: 115,200
  • LED indicators: red and green
  • Push-button start and stop: Yes
  • Battery life: 10 years typical at a 15 minute reading rate (ambienbt temp.)
  • Material: ABS plastic
  • Dimensions: 1.4 x 2.2 x 0.6 in. (3.6 x 5.6 x 16cm)
  • Operating environment: -40°Cã+80°C (-40°F to +176°F), 0ã95%RH non-condensing
  • Required interface package: IFC200
  • Weight: 0.0563 lb.

Order Info

Software, Manual and USB Interface Cable Package required (p/n 15-060-036).

Compliance

CE approved.

Specifications

Type Temperature and Humidity Datalogger
Data Memory 1,000,000/Channel
Calibration Digital Calibration
Alarm Types High/Low
Battery Type 3.6 V Lithium
Battery Life 10 Years
Certifications/Compliance CE
Interface IFC200, USB
Material ABS Plastic
Model RHTEMP101A
Requirements Software, Manual, USB Interface Cable Package Required (p/n 15-060-036)
Sensor Types Precision RTD Element (Temperature), Internal Semiconductor (Humidity)
For Use With (Application) For implementation of HACCP programs, HVAC, warehouse monitoring, museum monitoring, medical and pharmaceutical applications
Humidity Range 0 to 95%
Temperature Resolution 0.01°C
Temperature Range (Metric) -40°C to +80°C
Operating Temperature (English) -40°F to +176°F
Operating Temperature (Metric) -40°C to +80°C
Weight (English) 0.9 oz.
Weight (Metric) 24 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.