Learn More
MEDNA SCIENTIFIC INC Human IgG1 Poly-Specificity Control Antibody - 100ug
Supplier:  MEDNA SCIENTIFIC INC H1308100UG
Encompass
Human IgG1 Poly-Specificity Control Antibody is a recombinant antibody that recognizes Baculovirus (BV) particles. Non-specific binding of an antibody to Baculovirus particles is a measure of the poly-specificity of that antibody.  H1308 is developed to be used as a control for poly-specific binding in analytical and molecular applications. For more information, please visit mednabio.com.
Catalog No. NC2087553
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.