Learn More
Cayman Chemical Humn Hepatocyte Nuclear Fact
SDP
Supplier:  Cayman Chemical 4101096 WELL
Encompass
A 15-LO-2/PEBP1 and ferroptosis inhibitor selectively inhibits the peroxidation of 1-stearoyl-2-arachidonoyl-sn-glycero-3-PE by the 15-LO-2/PEBP1 complex over 15-LO-2 alone in cell-free assays at 0.25 UM inhibits RSL3-induced ferroptosis in primary human bronchial epithelial cells (IC50 84 nM) increases survival in a mouse model of total body irradiation at 25 mg/kg
Catalog No. 50-294-2527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.