Learn More
TARGETMOL CHEMICALS INC HUP30 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TNU0864
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HUP30 is a potent vasodilating agent. HUP30 can stimulate soluble guanylyl cyclase activate K+ channels and block extracellular Ca2+ influx. purity: 99%
Catalog No. 50-257-8868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.