Learn More
Genscript Corporation GRO beta/CXCL2 Human
Supplier:  Genscript Corporation Z0330425
Encompass_Preferred
Human GRO- GRO- (MIP-2 ) and GRO- (MIP-2 ) are products of three distinct nonallelichuman genes GRO- and GRO- share 90% and 86% amino acid sequence homology respectively with GRO All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors GRO expression is inducible by serum or PDGF and/or by a variety of inflammatory mediators such as IL-1 and TNF in monocytes fibroblasts melanocytes and epithelial cells In certain tumor cell lines GRO is expressed constitutively Similar to other alpha chemokines the three GRO proteins are potent neutrophil attractants and activators In addition these chemokines are also active toward basophils
Catalog No. 50-000-59396
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.