Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Hyaluronan Quantikine ELISA Kit

Catalog No. SHYAL0
Change view
Click to view available options
Quantity:
1 Pack (6 Plates)
One 96-well plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SHYAL0 1 Pack (6 Plates)
DHYAL0 One 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SHYAL0 Supplier R&D Systems™ Supplier No. SHYAL0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hyaluronan Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Hyaluronan Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).

Specifications

Assay Range 0.63 to 40 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity 35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples
For Use With (Application) ELISA
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Tissue Homogenates (50 uL),Serum (25 uL),EDTA Plasma (25 uL),Heparin Plasma (25 uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.