Learn More
TARGETMOL CHEMICALS INC CORNUSIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4S23265MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Cornuside (Comuside) has immunomodulatory activity . 2. Cornuside suppresses expression levels of cytokine-induced proinflammatory and adhesion molecules in the human endothelial cells. 3. Cornuside can treat myocardial I/R and protect the liver from CCl4-induced acute hepatotoxicity by reducing oxidative stress and suppressing inflammatory responses. 4. Cornuside has anti-inflammatory activity by downregulations of iNOS and COX-2 due to NF-(kappa)B inhibition as well as the negative regulation of ERK1/2 p38 and JNK1/2 phosphorylations in RAW 264.7 cells. 5. Cornuside has protective potential against cerebral ischemic injury may be due to the suppression of intracellular Ca(2+) elevation and caspase-3 activity and improvements in mitochondrial energy metabolism and antioxidant properties. purity: 99%
Catalog No. 50-257-9826
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.