Learn More
Selleck Chemical LLC Hyaluronidase S4424-5mg
SDP
Supplier:  Selleck Chemical LLC S44245MG
Encompass_Preferred
Hyaluronidase (Hyaluronate 4-glycanohydrolase Hyaluronoglucosaminidase Amphadase Hydase Vitrase) is a naturally occurring enzyme that degrades hyaluronic acid which constitutes an essential part of the extracellular matrix It depolymerizes hyaluronic acid by cleavage of glycosidic bonds and has been utilized in ophthalmic surgery
Catalog No. 50-311-0246
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.