Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hyaluronidase (Bovine)

Catalog No. 89948157 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-157 5000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-157 Supplier Abnova™ Supplier No. P5299
Add to Cart
Add to Cart

Used for Enzyme

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 55kDa
Name Hyaluronidase (Bovine)
Preparation Method Native protein chromatographically purified from Bovine Testes
Quantity 5000 Units
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name HYALURONIDASE
Assays Hyaluronidase Assay
Biological Activity ≥3,000 USP/NF units per mg dry weight. One Unit equals a U.S.P./National Formulary Unit and is referenced to a standard NF Hyaluronidase.
Species Bovine
Protein Tag None
Expression System Native protein chromatographically purified from Bovine Testes
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.