Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hyaluronidase (From Streptomyces hyalurolyticus)

Catalog No. MP08320421 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
4 x 100 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08320421 4 x 100 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08320421 Supplier MP Biomedicals™ Supplier No. 08320421
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hyaluronidase (From Streptomyces hyalurolyticus)

  • The hyaluronidase is purified from Streptomyces hyalurolyticus.
  • This enzyme belongs to the endo-beta-hexosaminidase and endo-eliminase type enzyme that produces oligo-saccharide (unsaturated tetraose and pentose) by hydrolyzing the beta-1,4-linkage of hyaluronic acid.
  • Hyaluronidase is stable in both high temperature and acid or alkaline range.
  • It has high substrate specificity and hydrolyzes hyaluronic acid only in mucopolysaccharide.
TRUSTED_SUSTAINABILITY

Specifications

Host Species Lyophilized, salt free
For Use With (Application) Cell Biology & Analysis
Content And Storage Store at 0-5C
Source Streptomyces hyalurolyticus
Quantity 4 x 100 U
Product Type Proteins, Enzymes & Peptides
Form Solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.