Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hyamine Solution, 0.008 Molar (0.008 Molar) (Benzethonium Chloride Solution), Reagents
Hyamine Solution, 0.008 Molar (0.008 Molar) (Benzethonium Chloride Solution), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$227.00 - $231.50
Specifications
Chemical Name or Material | Hyamine Solution, 0.008 Molar (0.008 Molar) (Benzethonium Chloride Solution) |
---|---|
Color | Colorless |
CAS | 7732-18-5, 121-54-0 |
Boiling Point | 100°C |
Synonym | Benzethonium chloride |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
01-335-060
|
Reagents Holdings Llc
CH1149004C |
4 L | Glass Bottle |
Each for $227.00
|
|
||||
01-335-059
|
Reagents Holdings Llc
CH1149004C1 |
4 L | Safety Coated Glass |
Each for $231.50
|
|
||||
Specifications
Hyamine Solution, 0.008 Molar (0.008 Molar) (Benzethonium Chloride Solution) | |
7732-18-5, 121-54-0 | |
Benzethonium chloride | |
1.001 | |
Non-sterile |
Colorless | |
100°C | |
Reagent | |
4 L | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title