Learn More
Thomas Scientific hybex Media Storage Bottle, 250mL with GL45 Yellow Cap
Supplier: Thomas Scientific 1189P26
Borosilicate 3.3 hybex glass Autoclavable caps (GL45) & drip rings Permanent enamel graduation markings Large area for bottle marking/identification Autoclavable, reusable and breakage resistant Five cap colors hybex™ reusable media bottles are manufactured from high quality, borosilicate 3.3 glass. With their superior chemical resistance, these bottles are ideal for storage of reagents, culture media, biological fluids and a variety of other aqueous and non-aqueous solutions. A low coefficient of thermal expansion (3.3), provides temperature resistance that is far superior to that of standard lab glass (or soda lime glass). Hybex glass can withstand temperatures from -70 to 500°C, permitting repeated use in freezers, microwaves, autoclaves and dry heat sterilization chambers. All hybex bottles have permanent, white enamel graduations for volume measurement and a white identification box for marking or coding. All hybex media bottles include a drip-free sealing ring as we
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.