Learn More
Thomas Scientific Filter Paper 601 Size 20X20cm
SDP
Supplier:  Thomas Scientific 0374G13
Encompass
Ahlstrom's qualitative grades of filter paper are, by definition, recommended for use in analytical methods, which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. Ahlstrom offers a complete range of low ash filter papers for general laboratory work that provides you with the correct combination of particle retention, filtration speed, loading capacity and wet-strength. All our qualitative grades consist of > 95% super refined a-cellulose and linter fibers. Key Features Quantitative analysis Low ash content 0.06% Untreated High wet-strength Consistent performance Wet-strengthened grades available Custom cut Bulk packaging Ahlstrom 601: 2.5µm – Medium Speed This grade covers a wide range of laboratory applications and is used for clarifying liquids and for rapid filtration of coarser precipitates
Catalog No. 50-145-8600
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.