Learn More
ATCC ATCC Hybridoma Clone 83A25 (HB-10392), Rat, Frozen Hybridoma
Supplier:  ATCC HB10392
Encompass
Hybridoma clone 83A25 is a rat-derived hybridoma supplied as a frozen living culture and deposited with ATCC as a patent deposit. The material is provided "AS IS" and users must follow the supplier's recovery and handling instructions; use may be restricted by patent claims.
  • Supplied as a frozen living culture for research use.
  • Rat-derived hybridoma that secretes a monoclonal antibody.
  • Store at temperatures below -130°C, preferably in liquid nitrogen vapor.
  • Viability warranted for 30 days from shipment when handled per supplier instructions.
  • May be subject to patent restrictions; licensing may be required for some uses.
Catalog No. NC3724490