Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC ATCC TIB-103 (T11D7e2) Hybridoma cell line, Mus musculus

Supplier:  ATCC TIB103

Encompass

Murine hybridoma cell line derived from mouse B cells fused with myeloma, used for immunology research and monoclonal antibody production. Cells grow as lymphoblast-like suspension cultures under standard mammalian tissue culture conditions and are maintained in Dulbecco's Modified Eagle's Medium supplemented with fetal bovine serum.

  • Species: Mus musculus (mouse).
  • Cell type: Hybridoma; B lymphocyte.
  • Morphology: Lymphoblast.
  • Recommended medium: Dulbecco's Modified Eagle's Medium (DMEM); add fetal bovine serum to 10% final concentration.
  • Culture conditions: 37 °C, 5% CO2 in air.
  • Applications: Immunology and monoclonal antibody production.

Catalog No. NC3417668


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.