Description
Key Features and Benefits:
- More accurate results
- Unique chemistry can effectively separate hydrophobic analytes (such as vitamins) from phospholipids, unlike competitive product chemistries
- Elimination of proteins and phospholipid-induced ion suppression
- Simultaneous removal of proteins and phospholipids
- Simple, standardized methodology, analogous to traditional protein precipitation
- Alternative to complex traditional SPE method development
- Reproducible, consistent performance reduces need for reprocessing
- Less instrument downtime and longer column life
- Decreased run times by eliminating the need for gradients to clean columns between samples
- High throughput processing that is automatable and compatible with common robotic systems
- Ready-to-use, no preconditioning required
Specifications
Specifications
| Bed Weight | 50 mg |
| For Use With (Application) | Food and Beverage |
| For Use With (Equipment) | INTEGRA |
| Volume (Metric) | 1.25 mL (Tip) |