Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ ActiPro Powdered Cell Culture Media

Catalog No. SH3103701 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3103701 5 L
SH3103708 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3103701 Supplier Cytiva Supplier No. SH31037.01
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

Formulated to provide high yields of recombinant proteins in bioprocesses using Chinese hamster ovary (CHO) cell lines.

HyClone ActiPro cell culture medium is available in both liquid and powder forms for cell culture bioprocesses. This chemically defined medium is intended for use in combination with HyClone ActiSM medium and Cell Boost 7a and Cell Boost 7b supplements.

ActiPro media and supplements are formulated for industrial production of recombinant proteins, including those requiring complex post-translational modifications and correct protein folding. In addition, this engineered ActiPro system of media and supplements can significantly reduce production process time.

  • Demonstrates high yields of recombinant protein
  • Does not contain hypoxanthine and thymidine (HAT supplement)
  • Chemically defined ADCF mAb media formulation
  • Production medium for batch or fed-batch fermentation processes
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.