Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ AdvanceSTEM™ Amniotic Epithelial Expansion Kit

Catalog No. SH3090002 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3090002 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Hyclone Cell Culture products now a part of GE Healthcare.
Catalog No. SH3090002 Supplier HyClone™ Supplier No. SH30900.02

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Hyclone Cell Culture products now a part of GE Healthcare.

Perform rapid and consistent expansion of amniotic epithelial stem cells with HyClone™ AdvanceSTEM™ Amniotic Epithelial Expansion Kit.

  • Includes a proprietary basal Amniotic Epithelial Expansion Medium, to which an aliquot of HyClone AdvanceSTEM Growth Supplement is added at the time of use. Basal Amniotic Epithelial Expansion Medium and Growth Supplement should be used together, but are available as separate components
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2° to 8°C away from light
Description HyClone AdvanceSTEM Amniotic Epithelial Growth Medium
Cell Type Human Amniotic Epithelial Cells (HAEC)
Form Liquid
Product Type Stem Cell Differentiation Media
Sterility Sterile-filtered
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.