Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Bovine Serum Albumin (BSA)

Catalog No. SH3057401 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3057401 10 g
SH3057402 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3057401 Supplier Cytiva Supplier No. SH30574.01

Support cell culture growth with HyClone™ Bovine Serum Albumin (BSA).

  • pH 7.0
  • Lyophilized powder
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage 2° to 8° C
Form Lyophilized Powder
Product Type BSA
Sterility Non-sterile
Endotoxin Level ≤ 10 EU/mg
pH 6.5 to 7.5
Purity or Quality Grade Cell Culture Grade
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.