Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ CDM4PERMAb cell culture media

Catalog No. SH3087102 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3087102 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3087102 Supplier Cytiva Supplier No. SH30871.02
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemically defined Animal-Derived Component-Free (ADCF) media for use with PER.C6 cells

  • Chemically defined and animal-derived component-free (ADCF) formulation
  • This high-performance cell culture medium has been developed through the Metabolic Pathway Design process to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 technology
  • The medium has been successfully tested in a variety of applications, including fed-batch bioreactor cultures
  • Designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured according to cGMP guidelines
  • TRUSTED_SUSTAINABILITY

    Specifications

    Form Liquid
    Sterility Sterile
    Sterilization Method 0.2 μm Sterile-filtered
    Quantity 1000 mL
    Format Bottle
    Content And Storage 2° to 8°C, away from light
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.