Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Cell Boost 2 Supplement

Catalog No. SH3111402 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3111402 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3111402 Supplier Cytiva Supplier No. SH31114.02
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supplements cell culture with amino acids and glucose and manufactured to meet cGMP manufacturing standards and QC specifications.

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 2 Supplement provides amino acids and glucose to support fed batch cultures in research and bioprocessing applications.

Available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

  • Designed to support modern manufacturing processes, Cell Boost 2 is chemically defined (CD), animal-derived component-free (ADCF), protein-free, and hydrolysate-free.
  • For convenience, this cell culture supplement is available as a ready-to-use liquid format in addition to a dry powder format.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C, away from light
Form Liquid
Sterility Non-sterile
Without Additives No Poloxamer 188, No Glutamine
Quantity 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.