Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Cell Boost 7b Supplement

Catalog No. SH3102707 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
0.1 L
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3102707 1 L
SH3102708 0.1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SH3102707 Supplier Cytiva Supplier No. SH31027.07
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplements cell culture with concentrated amino acids. Manufactured to meet cGMP manufacturing standards and QC specifications.

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines.

Cell Boost 7b is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

  • Designed to support modern manufacturing processes, Cell Boost 7b supplement is a concentrated solution of amino acids with an alkaline pH. Supplement is chemically defined, animal-derived component-free (ADCF), do not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Use Cell Boost 7a and 7b together to achieve high yields of mAbs and other proteins in CHO fed-batch processes
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No Poloxamer 188, No Insulin
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.