Learn More
Description
- Meets the requirements of most users
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Specifications
| Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein |
| Certifications/Compliance | Certificate of Suitability No. R1-CEP 2000-384 |
| Color | Amber |
| Content And Storage | Store at or Below -10°C |
| Species | Bovine |
| Age | Fetal |
| Country of Origin | Australia |
| Form | Serum |
| Format | Bottle |
| Product Type | Fetal Bovine Serum, Sera, Serum |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.