Learn More
Description
- HyClone Characterized Fetal Bovine Serum (FBS), Australian Origin is sourced from Australian abattoirs approved by the U.S. Department of Agriculture (USDA) or Australian Quarantine and Inspection Service (AQIS).
- The Australian methods of animal husbandry exhibit excellent animal nutrition and healthcare.
- Australia is an island continent, thus making animal disease and control management easier than in most areas of the world.
- Characterized FBS, Australian Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
- Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
Specifications
Specifications
| Additional Assays | Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG |
| Certifications/Compliance | USDA Regulations, AQIS |
| Color | Amber |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C |
| Species | Bovine |
| Age | Fetal |
| Country of Origin | Australia |
| Form | Frozen |
| Format | Bottle |
| Product Type | Fetal Bovine Serum |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.