Learn More
Description
- Enhance growth of cells
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 100 nm pore-size rated filters
Specifications
Specifications
| Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride |
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | United States |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Processing | Irradiated (30 to 45 doses) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.