Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)

Catalog No. SH300870445 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH300870445 1000 mL Irradiated, 30 to 45 doses
SH3008702 100 mL None
SH3008703 500 mL None
SH3008704 1000 mL None
SH3008703HI 500 mL Heat Inactivated
SH300870440 1000 mL Irradiated, 25 to 40 doses
SH300870340 500 mL Irradiated, 25 to 40 doses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. SH300870445 Supplier Cytiva Supplier No. SH30087.04IR3045
Only null left
Add to Cart
Add to Cart

Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.

  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Additive Iron and naturally derived components
Quantity 1000 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing Irradiated, 30 to 45 doses
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.