Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Cosmic Calf Serum (U.S. origin)

Catalog No. SH3008702HI Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3008702HI 100 mL
SH3008704HI 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3008702HI Supplier Cytiva Supplier No. SH30087.02HI
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.

HyClone Cosmic Calf serum is a high-performing bovine calf serum that has been fortified with iron and a proprietary blend of growth-promoting factors. The result is a naturally derived, high-quality engineered serum that has demonstrated excellent performance with a broad range of cell types. All components of Cosmic Calf serum are sourced in the USA and are traceable.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Product Type FBS Replacement Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Heat Inactivated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.