Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hyclone™ Donor Equine Serum

Catalog No. SH3007404 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 L
Processing:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3007404 1 L None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3007404 Supplier Cytiva Supplier No. SH30074.04
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carefully collected from selected U.S. donor herds that receive regular veterinary inspection and care.

Serum Collection:
  • From collection to final packaging
  • Every step is documented
  • Full traceability compliant with current Good Manufacturing Practices (cGMP)
Quick and Reliable Processing:
  • Extensive quality, performance, and analytical testing
  • Aseptically packaged in a Class 100 clean room condition
Quality Control Testing Standards:
  • Each lot of serum is tested to ensure quality and sterility
True Pool Processing:
  • Pooled after filtration and before final packaging
  • Consistent quality from the first bottle filled to the last
  • Increases consistency and minimizes variability with a lot of serum
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin
Content And Storage Store at -10°C or lower.
Species Equine
Age Adult
Country of Origin United States
Form Frozen
Format Bottle
Product Type Equine Serum
Quantity 1 L
Serum Treatment Sterile-filtered
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents
Heat Inactivated No
Product Line HyClone
Gamma-irradiated No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.