Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Donor Equine Serum (U.S. origin)

Catalog No. SH300740340 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 kGy)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH300740340 500 mL Irradiated (25 to 40 kGy)
SH3007402HI 100 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH300740340 Supplier Cytiva Supplier No. SH30074.03IR2540
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.

  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Content And Storage Store at or Below -10°C
    Species Equine
    Age Adult
    Country of Origin United States
    Form Frozen
    Format Bottle
    Product Type Equine Serum
    Quantity 500 mL
    Serum Treatment Gamma-irradiated
    Sterility Sterile-filtered
    Sterilization Method Sequential-filtered, 100 nm
    Processing Irradiated (25 to 40 kGy)
    Tested For Equine Herpesvirus, Equine Viral Arteritis, Bovine Virus Diarrhea, Rabies, Reovirus, Cytopathogenic Agnets (IBR), Hemabsorbing Agent (PI3), Equine Infectious Anemia
    Heat Inactivated No
    Product Line HyClone
    Shipping Condition Frozen
    Gamma-irradiated Yes
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.