Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Donor Equine Serum, US Origin

Catalog No. SH3744IR254 Shop All Cytiva Products
Change view
Click to view available options
Processing:
Irradiated (25 to 40 kGy)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Processing
SH3744IR254 Irradiated (25 to 40 kGy)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3744IR254 Supplier Cytiva Supplier No. SH30074.04IR2540
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).

  • Carefully collected from animals fed an enriched diet to ensure proper nutrition
  • To reduce lipid concentration, the equine herd is fasted and then the blood is collected in a special facility using aseptic techniques
  • Coggins tests for equine infectious anemia are performed on each horse
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or Below -10°C
Species Equine
Age >6 months
Country of Origin United States
Form Liquid
Format Bottle
Product Type Equine Serum, Sera, Serum
Quantity 1000 mL
Serum Treatment Gamma-irradiated
Sterility Sterile-filtered
Sterilization Method Sequential Triple-filtered, 100 nm
Processing Irradiated (25 to 40 kGy)
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Product Line HyClone
Shipping Condition Frozen
Gamma-irradiated Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.