Learn More
Description
- Filtered through 3 sequential 0.1 μm pore size filters
Specifications
Specifications
| Additional Assays | Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality |
| Certifications/Compliance | AQIS |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | Australia |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Processing | Irradiated (30 to 45 doses) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.