Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Catalog No. SH3008403HI Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3008403HI 500 mL Heat Inactivated
SH300840340 500 mL Irradiated (25 to 40 doses)
SH300840345 500 mL Irradiated (30 to 45 doses)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3008403HI Supplier Cytiva Supplier No. SH30084.03HI
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

  • Filtered through 3 sequential 0.1 μm pore size filters
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AQIS
Color Amber
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Species Bovine
Age Fetal
Country of Origin Australia
Form Frozen
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Heat-inactivated
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Grade Characterized
Processing Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤25 mg/dL
Product Line HyClone
Osmolality 280 to 360 mOsm/kg
pH 6.7 to 7.6
Protein Content 3 to 4.5 g/dL
USDA Safety Testing Included USDA Inspected
Gamma-irradiated No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.