Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)

Catalog No. SH3007001 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
None
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3007001 50 mL None
SH3007001HI 50 mL Heat Inactivated
SH3007002 100 mL None
SH3007002HI 100 mL Heat Inactivated
SH3007003 500 mL None
SH3007003HI 500 mL Heat Inactivated
SH300700340 500 mL Irradiated (25 to 40 doses)
SH300700345 500 mL Irradiated (30 to 45 doses)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. SH3007001 Supplier Cytiva Supplier No. SH30070.01
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

  • Maximize cell culture growth
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 40 nm pore-size rated filters
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays 1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 50 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 40 nm
Grade Defined
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Product Line HyClone
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.