Learn More
Description
Augment growth of cells with U.S.-sourced HyClone™ Dialyzed Fetal Bovine Serum (FBS), depleted of small molecules (less than 10,000 MW) and filtered through sequential 100nm (0.1μm) pore-size rated filters.
Specifications
Specifications
| Additional Assays | 1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C |
| Certifications/Compliance | EMEA Certified |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Description | Dialyzed Fetal Bovine Serum, US Origin, None |
| Country of Origin | United States |
| Quantity | 100 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Grade | Specialty filtered |
| Processing | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.