Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Bovine Serum, USDA Tested

Catalog No. p-7058019 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
None
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3091002 100 mL None
SH3091003 500 mL None
SH3091002HI 100 mL Heat Inactivated
SH3091003HI 500 mL Heat Inactivated
SH309100340 500 mL Irradiated (25 to 40 doses)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. SH3091002 Supplier Cytiva Supplier No. SH30910.02
Only null left
Add to Cart
Add to Cart

Sourced from countries recognized as being free of BSE and FMD by the USDA

Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.

Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Lactate Dehydrogenase, Osmolality, pH, Potassium, SGOT, SGPT, Sodium, Total Bilirubin, Total Protein
Certifications/Compliance USDA Regulations
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description USDA Tested Fetal Bovine Serum, Central American Sourced
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Characterized
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 25 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.