Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Clone I Serum

Catalog No. SH300800340 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH300800340 500 mL Irradiated (25 to 40 doses)
SH3008003HI 500 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH300800340 Supplier Cytiva Supplier No. SH30080.03IR2540
Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

HyClone™ FetalClone™ I is a fetal bovine serum alternative optimised for the growth of hybridomas with comparable IgG levels to those found in FBS.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Country of Origin United States
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated (25 to 40 doses)
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 20 mg/dL
IgG levels Typically less than 2 μg/mL
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.